Sodium 3-nitrobenzenesulfonate, hydrate ≥98%
Supplier: BeanTown Chemical
|
Warning
|
Synonyms:
Sodium 3-nitrobenzenesulphonate, 3-Nitrobenzenesulphonic acid, sodium salt
216470-100G
216470-2.5KG
216470-500G
BT216470-100GEA
25
USD
BT216470-100G
BT216470-2.5KG
BT216470-500G
Sodium 3-nitrobenzenesulfonate, hydrate ≥98%
Sodium-3-nitrobenzenesulphonate
CAS: 127-68-4; EC No: 204-857-3; MDL No: MFCD00007490; RTECS: DB7195000
Powder; Molecular Formula: C6H4NNaO5S·xH2O; MW: 225.15
Melting Point: 350°; Flash point: 100°C (212°F)
Moisture Sensitive
- RTECS: DB7195000
- Moisture Sensitive
Formula:
C₆H₄NNaO₅S MW: 225.16 g/mol Melting Pt: 350 °C Flash Pt: 100 °C (212 °F) |
MDL Number:
MFCD00007490 CAS Number: 127-68-4 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...