(S)-(-)-4-Benzyl-2-oxazolidinone ≥99%
Supplier: BeanTown Chemical
Synonyms:
(S)-4-Benzyl-2-oxazolidinone, (S)-4-Benzyloxazolidin-2-one
216450-1G
216450-25G
216450-5G
BT216450-1GEA
33
USD
BT216450-1G
BT216450-25G
BT216450-5G
(S)-(-)-4-Benzyl-2-oxazolidinone ≥99%
(S)-(-)-4-Benzyl-2-oxazolidinone
CAS: 90719-32-7; MDL No: MFCD00064496
Crystalline; Molecular Formula: C10H11NO2; MW: 177.20
Melting Point: 86-88°
Optical Rotation: [α]20/D -63°, c = 1 in chloroform
Hygroscopic
- Optical rotation: [α]20/D -63°, c = 1 in chloroform
- Hygroscopic
- Crystalline
Formula:
C₁₀H₁₁NO₂ Melting Pt: 86…88 °C |
MDL Number:
MFCD00064496 CAS Number: 90719-32-7 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...