2-Amino-5-bromo-3-nitropyridine ≥97%

Supplier: BeanTown Chemical

Synonyms: 5-Bromo-3-nitropyridin-2-amine, 5-Bromo-3-nitro-2-pyridylamine

216445-1G 216445-25G 216445-5G
BT216445-1GEA 33 USD
BT216445-1G BT216445-25G BT216445-5G
2-Amino-5-bromo-3-nitropyridine ≥97%
2-Amino-5-bromo-3-nitropyridine

CAS: 6945-68-2; MDL No: MFCD00047441 Crystalline/Powder; Molecular Formula: C5H4BrN3O2 ; MW: 218.01 Melting Point: 205-208°


  • Crystalline/Powder

Formula: C₅H₄BrN₃O₂
MW: 218.01 g/mol
Melting Pt: 205…208 °C
MDL Number: MFCD00047441
CAS Number: 6945-68-2

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR