Manganese(II) phthalocyanine

Supplier: BeanTown Chemical
Warning

216440-1G 216440-5G
BT216440-1GEA 44 USD
BT216440-1G BT216440-5G
Manganese(II) phthalocyanine
Manganese(II) phthalocyanine

CAS: 14325-24-7; MDL No: MFCD00049821 Crystalline; Molecular Formula: C32H16MnN8 ; MW: 567.47


  • Crystalline

Formula: C₃₂H₁₆MnN₈
MW: 567.46 g/mol
MDL Number: MFCD00049821
CAS Number: 14325-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR