(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone ≥99%

Supplier: BeanTown Chemical

216415-1G 216415-5G
BT216415-1GEA 81 USD
BT216415-1G BT216415-5G
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone ≥99%
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone

CAS: 66673-40-3; MDL No: MFCD00077791 Powder; Molecular Formula: C5H9NO2; MW: 115.13 Melting Point: 83-85°; Boiling Point: 147-149°/0.06 mmHg Optical Rotation: [α]20/D −31°, c = 5 in ethanol


  • Optical rotation: [α]20/D −31°, c = 5 in ethanol
  • Powder

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 147…149 °C/0.06 mmHg
Melting Pt: 83…85 °C
MDL Number: MFCD00077791
CAS Number: 66673-40-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR