Benzoic acid ≥99.5% ACS
Supplier: BeanTown Chemical
216390-100G
216390-25G
216390-500G
BT216390-100GEA
51.81
USD
BT216390-100G
BT216390-25G
BT216390-500G
Benzoic acid ≥99.5% ACS
Benzoic acid
CAS: 65-85-0; EC No: 200-618-2; MDL No: MFCD00002398; RTECS: DG0875000
Crystalline; Linear Formula: C6H5CO2H; MW: 122.12
Melting Point: 121-125°; Boiling Point: 249°; Flash point: 121°C (250°F)
- RTECS: DG0875000
- Crystalline
Formula:
C₇H₆O₂ MW: 122.12 g/mol Boiling Pt: 249 °C Melting Pt: 121…125 °C Flash Pt: 121 °C (250 °F) |
MDL Number:
MFCD00002398 CAS Number: 65-85-0 EINECS: 200-618-2 |
Specification Test Results
Assay | 99.5% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...