Methyl-2,2-dimethylphenylacetate
Supplier: BeanTown Chemical
Synonyms:
Methyl 2-methyl-2-phenylpropanoate
216375-1G
216375-5G
BT216375-1GEA
123
USD
BT216375-1G
BT216375-5G
Methyl-2,2-dimethylphenylacetate
Methyl-2,2-dimethylphenylacetate
CAS: 57625-74-8
Liquid; Molecular Formula: C11H14O2; MW: 178.23
- Liquid
Formula:
C₁₁H₁₄O₂ MW: 178.231 g/mol |
MDL Number:
MFCD01846451 CAS Number: 57625-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...