Methylene blue hydrate
Supplier: BeanTown Chemical
|
Warning
|
Synonyms:
Methylthioninium chloride hydrate, Basic Blue 9, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride hydrate, C.I. 52015, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride xhydrate
216350-100G
216350-25G
216350-500G
BT216350-100GEA
68
USD
BT216350-100G
BT216350-25G
BT216350-500G
Methylene blue hydrate
Methylene blue hydrate
CAS: 122965-43-9; EC No: 200-515-2; MDL No: MFCD00150006; RTECS: SO5600000
Powder; Molecular Formula: C16H18ClN3S·xH2O; MW: 319.85 (anhydrous)
Melting Point: 180° (decomposes)
Formula:
C₁₆H₁₈ClN₃S·xH₂O Melting Pt: 180 °C (dec.) |
MDL Number:
MFCD00150006 CAS Number: 122965-43-9 EINECS: 200-515-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...