Orcein

Supplier: BeanTown Chemical

Synonyms: Orcein (from synthetic), Natural Red 28

216340-1G 216340-5G
BT216340-1GEA 34 USD
BT216340-1G BT216340-5G
Orcein
Orcein

CAS: 1400-62-0; EC No: 215-750-6; MDL No: MFCD00062310 Powder


  • Powder

MDL Number: MFCD00062310
CAS Number: 1400-62-0
EINECS: 215-750-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR