Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate

Supplier: BeanTown Chemical

216330-100G 216330-500G
BT216330-100GEA 32 USD
BT216330-100G BT216330-500G
Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate
Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate

CAS: 123409-01-8; EC No: 225-962-0; MDL No: MFCD00149263 Solid; Linear Formula: C10H5(SO3Na)3·xH2O ; Molecular Formula: C10H5Na3O9S3·xH2O ; MW: 434.31 (anhydrous)


  • Solid

Formula: C₁₀H₅Na₃O₉S₃·xH₂O
MDL Number: MFCD00149263
CAS Number: 123409-01-8
EINECS: 225-962-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR