Succinic acid ≥99.0% ACS
Supplier: BeanTown Chemical
Synonyms:
1,4-Butanedioic acid
216325-2KG
216325-500G
BT216325-2KGEA
144.81
USD
BT216325-2KG
BT216325-500G
Succinic acid ≥99.0% ACS
Succinic acid
CAS: 110-15-6; EC No: 203-740-4; MDL No: MFCD00002789; RTECS: WM4900000
Crystalline ; Linear Formula: HOOCCH2CH2COOH; Molecular Formula: C4H6O4; MW: 118.09
Melting Point: 184-186°; Boiling Point: 235°; Flash point: 206°C (402°F)
Density (g/mL): 1.564
- RTECS: WM4900000
- Crystalline
Formula:
HOOCCH₂CH₂COOH MW: 118.09 g/mol Boiling Pt: 235 °C Melting Pt: 184…186 °C Density: 1.564 g/mL Flash Pt: 206 °C (402 °F) |
MDL Number:
MFCD00002789 CAS Number: 110-15-6 EINECS: 203-740-4 UN: 3261 ADR: 8,III |
Specification Test Results
Assay | 99.0% min |
Melting range | 185.0-187.0°C |
Insoluble matter | ≤0.01% |
Residue after ignition | ≤0.02% |
Chloride (Cl) | ≤0.001% |
Phosphate (PO4) | ≤0.001% |
Sulfate (SO4) | ≤0.003% |
Nitrogen compounds (as N) | ≤0.001% |
Heavy metals (as Pb) | ≤5ppm |
Iron (Fe) | ≤5ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...