7-Bromoindole ≥98%

Supplier: BeanTown Chemical

Synonyms: 7-Bromo-1H-indole

216320-1G 216320-250MG 216320-5G
BT216320-1GEA 292 USD
BT216320-1G BT216320-250MG BT216320-5G
7-Bromoindole ≥98%
7-Bromoindole

CAS: 51417-51-7; MDL No: MFCD00799492 Powder; Molecular Formula: C8H6BrN; MW: 196.04 Melting Point: 41-44° Light Sensitive


  • Light Sensitive
  • Powder

Formula: C₈H₆BrN
MW: 196.05 g/mol
Melting Pt: 41…44 °C
MDL Number: MFCD00799492
CAS Number: 51417-51-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR