Nicotinic acid (Niacin) ≥99%

Supplier: BeanTown Chemical

216300-1KG 216300-250G 216300-5KG
BT216300-1KGEA 88.56 USD
BT216300-1KG BT216300-250G BT216300-5KG
Nicotinic acid (Niacin) ≥99%
Nicotinic acid (Niacin)

CAS: 59-67-6; EC No: 200-441-0; MDL No: MFCD00006391; RTECS: QT0525000 Powder; Molecular Formula: C6H5NO2; MW: 123.11 Melting Point: 236-239°; Flash point: 193°C (379°F) Density (g/mL): 1.467 Light Sensitive

Formula: C₆H₅NO₂
MW: 123.11 g/mol
MDL Number: MFCD00006391
CAS Number: 59-67-6

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR