Retinoic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
all-trans-Retinoic acid, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid, ATRA, Vitamin A acid, Tretinoin
216220-1G
216220-250MG
BT216220-1GEA
160.9
USD
BT216220-1G
BT216220-250MG
Retinoic acid ≥98%
Retinoic acid
CAS: 302-79-4; EC No: 206-129-0; MDL No: MFCD00001551; RTECS: VH6475000
Crystalline/Powder; Molecular Formula: C20H28O2; MW: 300.44
Melting Point: 180-181°
Light Sensitive
- RTECS: VH6475000
- Light Sensitive
- Crystalline/Powder
Formula:
C₂₀H₂₈O₂ MW: 300.44 g/mol Melting Pt: 180…181 °C |
MDL Number:
MFCD00001551 CAS Number: 302-79-4 EINECS: 206-129-0 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...