Gum arabic (total ash max 4%)
Supplier: BeanTown Chemical
|
Warning
|
Synonyms:
Acacia, Acacia gum
216190-2KG
216190-500G
BT216190-2KGEA
223
USD
BT216190-2KG
BT216190-500G
Gum arabic (total ash max 4%)
Gum arabic
CAS: 9000-01-5; EC No: 232-519-5; MDL No: MFCD00081264
Powder
Density:
0.64 g/cm³ (5.3408 lbs/gal) |
MDL Number:
MFCD00081264 CAS Number: 9000-01-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...