Calcium hexaboride ≥99.5%

Supplier: BeanTown Chemical

216185-100G 216185-25G 216185-5G
BT216185-100GEA 371 USD
BT216185-100G BT216185-25G BT216185-5G
Calcium hexaboride ≥99.5%
Calcium hexaboride

CAS: 12007-99-7; EC No: 234-525-3; MDL No: MFCD00148904 -325 Mesh Powder; Linear Formula: CaB6 ; MW: 104.94 Density (g/mL): 2.3


  • -325 Mesh Powder

Formula: B₆Ca
Density: 2.3 g/mL
MDL Number: MFCD00148904
CAS Number: 12007-99-7
EINECS: 234-525-3

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR