2-(Dicyclohexylphosphino)-2'-methylbiphenyl ≥98%
Supplier: BeanTown Chemical
Synonyms:
Dicyclohexyl(2'-methyl-[1,1'-biphenyl]-2-yl)phosphine, 2-Dicyclohexylphosphino-2'-methyl-)-1,1'-biphenyl, MePhos, 2-Dicyclohexylphosphino-2'-methylbiphenyl
216180-1G
216180-25G
216180-5G
BT216180-1GEA
166
USD
BT216180-1G
BT216180-25G
BT216180-5G
2-(Dicyclohexylphosphino)-2'-methylbiphenyl ≥98%
2-(Dicyclohexylphosphino)-2'-methylbiphenyl
CAS: 251320-86-2; MDL No: MFCD03094577
Crystalline/Powder; Molecular Formula: C25H33P; MW: 364.50
Melting Point: 105-109°
Air Sensitive
- Air Sensitive
- Crystalline/Powder
Formula:
C₂₅H₃₃P MW: 364.513 g/mol Melting Pt: 105…109 °C |
MDL Number:
MFCD03094577 CAS Number: 251320-86-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...