Zinc 2-ethylhexanoate

Supplier: BeanTown Chemical

Synonyms: Zinc(II) 2-ethylhexanoate

216140-100G 216140-500G
BT216140-100GEA 45.2 USD
BT216140-100G BT216140-500G
Zinc 2-ethylhexanoate
Zinc 2-ethylhexanoate

CAS: 136-53-8; EC No: 205-251-1; MDL No: MFCD00053316 Liquid; Linear Formula: [CH3(CH2)3CH(C2H5)CO2]2Zn; Molecular Formula: C16H30O4Zn; MW: 351.79


  • Liquid

Formula: C₁₆H₃₀O₄Zn
MW: 351.792 g/mol
MDL Number: MFCD00053316
CAS Number: 136-53-8
EINECS: 205-251-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR