Polypropylene glycol, M.W. ∼ 3,000

Supplier: BeanTown Chemical

Synonyms: Poly(propylene glycol), Poly(propylene oxide), PPG

216095-250G 216095-50G
BT216095-250GEA 91 USD
BT216095-250G BT216095-50G
Polypropylene glycol, M.W. ∼ 3,000
Polypropylene glycol

CAS: 25322-69-4; MDL No: MFCD00084446 Liquid; Linear Formula: H[OCH(CH3)CH2]nOH; MW ≈3,000 Density (g/mL): 1.006


  • Liquid

Formula: H(OCH₂CH₂CH₂)nOH
Density: 1.006 g/mL
MDL Number: MFCD00084446
CAS Number: 25322-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR