Triethyl methanetricarboxylate ≥98%

Supplier: BeanTown Chemical

216075-25G 216075-5G
BT216075-25GEA 69 USD
BT216075-25G BT216075-5G
Triethyl methanetricarboxylate ≥98%
Triethyl methanetricarboxylate

CAS: 6279-86-3; EC No: 228-477-2; MDL No: MFCD00009150 Liquid; Linear Formula: CH(COOCH2CH3)3; Molecular Formula: C10H16O6; MW: 232.23 Melting Point: 25-26°; Boiling Point: 253°; Flash point: 110°C (230°F) Density (g/mL): 1.095; Refractive Index: 1.424


  • Refractive index: 1.424
  • Liquid

Formula: C₁₀H₁₆O₆
MW: 232.23 g/mol
Boiling Pt: 253 °C
Melting Pt: 25…26 °C
Density: 1.095 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00009150
CAS Number: 6279-86-3
EINECS: 228-477-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR