(R)-β-Phenylalaninol ≥97%
Supplier: BeanTown Chemical
Synonyms:
(R)-3-Amino-3-phenylpropan-1-ol, (R)-1-Phenyl-3-propanolamine, (R)-3-Amino-3-phenyl-1-propanol
216070-1G
216070-5G
BT216070-1GEA
216
USD
BT216070-1G
BT216070-5G
(R)-β-Phenylalaninol ≥97%
(R)-β-Phenylalaninol
CAS: 170564-98-4; MDL No: MFCD01311791
Solid; Molecular Formula: C9H13NO; MW: 151.21
- Solid
Formula:
C₉H₁₃NO MW: 151.2056 g/mol |
MDL Number:
MFCD01311791 CAS Number: 170564-98-4 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...