5-Chlorotryptamine hydrochloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
5-Chloro-3-(2-aminoethyl)indole Hydrochloride
216060-1G
216060-5G
BT216060-1GEA
176
USD
BT216060-1G
BT216060-5G
5-Chlorotryptamine hydrochloride ≥98%
5-Chlorotryptamine hydrochloride
CAS: 942-26-7; EC No: 213-387-8; MDL No: MFCD00051990
Powder; Molecular Formula: C10H11ClN2·HCl; MW: 231.13
Melting Point: 290° (decomposes)
- Powder
Formula:
C₁₀H₁₂Cl₂N₂ MW: 231.12 g/mol Melting Pt: 290 °C (dec.) |
MDL Number:
MFCD00051990 CAS Number: 942-26-7 EINECS: 213-387-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...