Chloramphenicol ≥99%

Supplier: BeanTown Chemical

Synonyms: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol

216035-100G 216035-25G
BT216035-100GEA 154.5 USD
BT216035-100G BT216035-25G
Chloramphenicol ≥99%
Chloramphenicol

  • Powder

Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
Melting Pt: 148…150 °C
MDL Number: MFCD00078159
CAS Number: 56-75-7
Merck Index: 14,2077

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR