Magnesium hydroxide ≥95%

Supplier: BeanTown Chemical

215970-100G 215970-2.5KG 215970-500G
BT215970-100GEA 26.08 USD
BT215970-100G BT215970-2.5KG BT215970-500G
Magnesium hydroxide ≥95%
Magnesium hydroxide

CAS: 1309-42-8; EC No: 215-170-3; MDL No: MFCD00011104 Powder; Linear Formula: Mg(OH)2; MW: 58.33 Melting Point: 350° (decomposes) Density (g/mL): 2.36 Air Sensitive


  • Air Sensitive
  • Powder

Formula: Mg(OH)₂
MW: 58.32 g/mol
Melting Pt: 350 °C (dec.)
Density: 2.36 g/mL
MDL Number: MFCD00011104
CAS Number: 1309-42-8
EINECS: 215-170-3
Merck Index: 13,05693

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR