Disodium terephthalate ≥99%

Supplier: BeanTown Chemical

215960-100G 215960-25G 215960-5G
BT215960-100GEA 100.03 USD
BT215960-100G BT215960-25G BT215960-5G
Disodium terephthalate ≥99%
Disodium terephthalate

CAS: 10028-70-3; EC No: 233-075-5; MDL No: MFCD00013137 Powder; Linear Formula: C6H4(COONa)2; MW: 210.10


  • Powder

Formula: C₈H₄Na₂O₄
MDL Number: MFCD00013137
CAS Number: 10028-70-3
EINECS: 233-075-5

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR