Phloroglucinol, anhydrous ≥98%
Supplier: BeanTown Chemical
Synonyms:
1,3,5-Trihydroxybenzene
215910-250G
215910-50G
BT215910-250GEA
217
USD
BT215910-250G
BT215910-50G
Phloroglucinol, anhydrous ≥98%
Phloroglucinol
CAS: 108-73-6; EC No: 203-611-2; MDL No: MFCD00002286; RTECS: SY1050000
Solid; Linear Formula: C6H3(OH)3; MW: 126.11
Melting Point: 215-220°
Hygroscopic , Light Sensitive
- RTECS: SY1050000
- Hygroscopic, Light Sensitive
- Solid
Formula:
C₆H₆O₃ MW: 126.11 g/mol Melting Pt: 215…220 °C |
MDL Number:
MFCD00002286 CAS Number: 108-73-6 EINECS: 203-611-2 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...