Pyrazine-2,3-dicarboxylic acid ≥98%
Supplier: BeanTown Chemical
215900-100G
215900-25G
215900-5G
BT215900-100GEA
288
USD
BT215900-100G
BT215900-25G
BT215900-5G
Pyrazine-2,3-dicarboxylic acid ≥98%
Pyrazine-2,3-dicarboxylic acid
CAS: 89-01-0; EC No: 201-875-3; MDL No: MFCD00006131; RTECS: UQ2500000
Powder; Molecular Formula: C6H4N2O4 ; MW: 168.11
Melting Point: 188° (decomposes)
- RTECS: UQ2500000
- Powder
Formula:
C₆H₄N₂O₄ Melting Pt: 188 °C (dec.) |
MDL Number:
MFCD00006131 CAS Number: 89-01-0 EINECS: 201-875-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...