Dimethyl methylphosphonate ≥97%
Supplier: BeanTown Chemical
215875-100G
215875-500G
BT215875-100GEA
27.83
USD
BT215875-100G
BT215875-500G
Dimethyl methylphosphonate ≥97%
Dimethyl methylphosphonate
CAS: 756-79-6; EC No: 212-052-3; MDL No: MFCD00008349; RTECS: SZ9120000
Liquid; Linear Formula: CH3P(O)(OCH3)2; Molecular Formula: C3H9O3P; MW: 124.08
Melting Point: 181°; Flash point: 69°C (156°F)
Density (g/mL): 1.145; Refractive Index: 1.413
- RTECS: SZ9120000
- Refractive index: 1.413
- Liquid
Formula:
C₃H₉O₃P MW: 124.08 g/mol Melting Pt: 181 °C Density: 1.145 g/mL Flash Pt: 69 °C (156 °F) |
MDL Number:
MFCD00008349 CAS Number: 756-79-6 EINECS: 212-052-3 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...