Strontium titanate

Supplier: BeanTown Chemical

215870-100G 215870-500G
BT215870-100GEA 211 USD
BT215870-100G BT215870-500G
Strontium titanate
Strontium titanate

CAS: 12060-59-2; EC No: 235-044-1; MDL No: MFCD00049554 0.05-0.15 Micron APS Powder; Linear Formula: SrTiO3; MW: 183.50 Melting Point: 2060° Density (g/mL): 4.810


  • 0.05-0.15 Micron APS Powder

Formula: O₃SrTi
Melting Pt: 2060 °C
Density: 4.810 g/mL
MDL Number: MFCD00049554
CAS Number: 12060-59-2
EINECS: 235-044-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR