4-Amino-N,N-dimethylbenzamide ≥97%
Supplier: BeanTown Chemical
215860-1G
215860-5G
BT215860-1GEA
206
USD
BT215860-1G
BT215860-5G
4-Amino-N,N-dimethylbenzamide ≥97%
4-Amino-N,N-dimethylbenzamide
CAS: 6331-71-1; MDL No: MFCD00523648
Solid; Molecular Formula: C9H12N2O; MW: 164.21
Melting Point: 151-153°
- Solid
Formula:
C₉H₁₂N₂O Melting Pt: 151…153 °C |
MDL Number:
MFCD00523648 CAS Number: 6331-71-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...