Trimethylsulphoxonium iodide ≥98%
Supplier: BeanTown Chemical
Synonyms:
Trimethyloxosulphonium iodide
215830-100G
215830-25G
BT215830-100GEA
45
USD
BT215830-100G
BT215830-25G
Trimethylsulphoxonium iodide ≥98%
Trimethylsulphoxonium iodide
CAS: 1774-47-6; EC No: 217-204-2; MDL No: MFCD00011899; RTECS: WS3585000
Solid; Linear Formula: (CH3)2SIO; MW: 220.07
Melting Point: 208-212° (decomposes)
Light Sensitive
- RTECS: WS3585000
- Light Sensitive
- Solid
Formula:
C₃H₉IOS MW: 220.07 g/mol Melting Pt: 208…212 °C (dec.) |
MDL Number:
MFCD00011899 CAS Number: 1774-47-6 EINECS: 217-204-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...