Indium phosphide ≥99.99% (trace metals basis), polycrystalline lumps 2-8 mm (0.08-0.3 in)
Supplier: BeanTown Chemical
Synonyms:
Indium(III) phosphide, Indium phosphide
215775-1G
215775-25G
215775-5G
BT215775-1GEA
92
USD
BT215775-1G
BT215775-25G
BT215775-5G
Indium phosphide ≥99.99% (trace metals basis), polycrystalline lumps 2-8 mm (0.08-0.3 in)
Indium phosphide
CAS: 22398-80-7; EC No: 244-959-5; MDL No: MFCD00016153; RTECS: NL1800000
Pieces; Molecular Formula: InP; MW: 145.79
- RTECS: NL1800000
- Pieces
Formula:
InP MW: 145.79 g/mol |
MDL Number:
MFCD00016153 CAS Number: 22398-80-7 EINECS: 244-959-5 |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...