2-Hydroxypyrimidine hydrochloride ≥98%

Supplier: BeanTown Chemical

215740-100G 215740-25G 215740-500G
BT215740-100GEA 161 USD
BT215740-100G BT215740-25G BT215740-500G
2-Hydroxypyrimidine hydrochloride ≥98%
2-Hydroxypyrimidine hydrochloride

CAS: 38353-09-2; EC No: 253-897-8; MDL No: MFCD00012781 Crystalline; Molecular Formula: C4H4N2O·HCl; MW: 132.55 Melting Point: 200-205° (decomposes) Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₄H₄N₂O
Melting Pt: 200…205 °C (dec.)
MDL Number: MFCD00012781
CAS Number: 38353-09-2
EINECS: 253-897-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR