4-Formylbenzoic acid ≥98%

Supplier: BeanTown Chemical

215715-10G 215715-250G 215715-50G
BT215715-10GEA 53 USD
BT215715-10G BT215715-250G BT215715-50G
4-Formylbenzoic acid ≥98%
4-Formylbenzoic acid
Formula: C₈H₆O₃
Melting Pt: 247 °C
MDL Number: MFCD00006951
CAS Number: 619-66-9
EINECS: 210-607-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR