Methacrylamide ≥98%
Supplier: BeanTown Chemical
215705-100G
215705-500G
BT215705-100GEA
21.6
USD
BT215705-100G
BT215705-500G
Methacrylamide ≥98%
Methacrylamide
CAS: 79-39-0; EC No: 201-202-3; MDL No: MFCD00008018; RTECS: UC6475000
Crystalline; Linear Formula: H2C=C(CH3)CONH2; Molecular Formula: C4H7NO; MW: 85.11
Melting Point: 107-112°; Boiling Point: 215°
Density (g/mL): 1.115
- RTECS: UC6475000
- Crystalline
Formula:
C₄H₇NO MW: 85.11 g/mol Boiling Pt: 215 °C Melting Pt: 107…112 °C Density: 1.115 g/mL |
MDL Number:
MFCD00008018 CAS Number: 79-39-0 EINECS: 201-202-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...