Bis(cyclopentadienyl)titanium dichloride ≥97%

Supplier: BeanTown Chemical

215680-10G 215680-250G 215680-50G
BT215680-10GEA 44 USD
BT215680-10G BT215680-250G BT215680-50G
Bis(cyclopentadienyl)titanium dichloride ≥97%
Bis(cyclopentadienyl)titanium dichloride

CAS: 1271-19-8; EC No: 215-035-9; MDL No: MFCD00003723; RTECS: XR2050000 Powder; Molecular Formula: C10H10Cl2Ti; MW: 248.96 Melting Point: 260-280° (decomposes) Density (g/mL): 1.6 Air Sensitive, Moisture Sensitive


  • RTECS: XR2050000
  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: C₁₀H₁₀Cl₂Ti
Melting Pt: 260…280 °C (dec.)
Density: 1.6 g/mL
MDL Number: MFCD00003723
CAS Number: 1271-19-8
EINECS: 215-035-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR