Zinc D-gluconate hydrate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Gluconic acid Zzinc(II) salt hydrate, Zinc(II) gluconate hydrate, Zinc gluconate hydrate
215635-100G
215635-500G
BT215635-100GEA
57.4
USD
BT215635-100G
BT215635-500G
Zinc D-gluconate hydrate ≥97%
Zinc D-gluconate hydrate
CAS: 4468-02-4; EC No: 224-736-9; MDL No: MFCD00868110; RTECS: ZH3750000
Solid; Linear Formula: Zn(C6H11O7)2·xH2O; MW: 455.68 (anhydrous)
- RTECS: ZH3750000
- Solid
Formula:
C₁₂H₂₂O₁₄Zn·xH₂O MW: 455.69 g/mol (anhy) |
MDL Number:
MFCD00868110 CAS Number: 4468-02-4 EINECS: 224-736-9 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...