Titanium(IV) oxide rutile ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Pigment White 6, Rutile, Titanium dioxide rutile
215615-100G
215615-25G
BT215615-100GEA
172
USD
BT215615-100G
BT215615-25G
Titanium(IV) oxide rutile ≥99.9%
Titanium(IV) oxide rutile
CAS: 1317-80-2; EC No: 215-282-2; MDL No: MFCD00011269; RTECS: XR2275000
3-6mm Sintered Pieces; Linear Formula: TiO2; MW: 79.90
Melting Point: 1830-1850°; Boiling Point: 2500-3000°
Density (g/mL): 4.23
- RTECS: XR2275000
- 3-6mm Sintered Pieces
Formula:
O₂Ti MW: 79.88 g/mol Boiling Pt: 2500…3000 °C Melting Pt: 1855 Density: 4.23 g/mL |
MDL Number:
MFCD00011269 CAS Number: 1317-80-2 EINECS: 215-282-2 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...