n-Dodecane ≥99%
Supplier: BeanTown Chemical
Synonyms:
Dodecane
215610-100ML
215610-2.5L
215610-500ML
BT215610-100MLEA
70.47
USD
BT215610-100ML
BT215610-2.5L
BT215610-500ML
n-Dodecane ≥99%
n-Dodecane
CAS: 112-40-3; EC No: 203-967-9; MDL No: MFCD00008969; RTECS: JR2125000
Liquid; Linear Formula: CH3(CH2)10CH3; Molecular Formula: C12H26; MW: 170.34
Melting Point: -10°; Boiling Point: 215-217°; Flash point: 71°C (160°F)
Density (g/mL): 0.749; Refractive Index: 1.421
- RTECS: JR2125000
- Refractive index: 1.421
- Liquid
Formula:
H₃C(CH₂)₁₀CH₃ MW: 170.34 g/mol Boiling Pt: 215…217 °C Melting Pt: –10 °C Density: 0.749 g/mL Flash Pt: 71 °C (160 °F) |
MDL Number:
MFCD00008969 CAS Number: 112-40-3 EINECS: 203-967-9 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...