2,2'-O-Cyclocytidine hydrochloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Ancitabine Hydrochloride, Cyclocytidine hydrochloride, (2R,3R,3aS,9aR)-2-(Hydroxymethyl)-6-imino-3,3a,6,9a-tetrahydro-2H-furo[2',3':4,5]oxazolo[3,2-a]pyrimidin-3-ol hydrochloride, 2,2'-Anhydro-(1-beta-D-arabinofuranosyl)cytosine Hydrochloride
215600-1G
215600-5G
BT215600-1GEA
42
USD
BT215600-1G
BT215600-5G
2,2'-O-Cyclocytidine hydrochloride ≥98%
2,2'-O-Cyclocytidine hydrochloride
CAS: 10212-25-6; EC No: 233-515-6; MDL No: MFCD00012636; RTECS: LV2615000
Crystalline/Powder; Molecular Formula: C9H12ClN3O4; MW: 261.66
- RTECS: LV2615000
- Crystalline/Powder
Formula:
C₉H₁₁N₃O₄ MW: 261.66 g/mol |
MDL Number:
MFCD00012636 CAS Number: 10212-25-6 EINECS: 233-515-6 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...