Niobium ≥99.8% (trace metals basis), foil, Thickness 2.0 mm
Supplier: BeanTown Chemical
215595-100X200
215595-50X100
BT215595-100X2EA
1065
USD
BT215595-100X2
BT215595-50X10
Niobium ≥99.8% (trace metals basis), foil, Thickness 2.0 mm
Niobium
CAS: 7440-03-1; EC No: 231-113-5; MDL No: MFCD00011126
Powder; Molecular Formula: Nb; MW: 92.91
Melting Point: 2468°; Boiling Point: 4742°
Density (g/mL): 8.57
- Powder
Formula:
Nb Boiling Pt: 4742 °C Melting Pt: 2468 °C Density: 8.57 g/mL |
MDL Number:
MFCD00011126 CAS Number: 7440-03-1 EINECS: 231-113-5 |
Specification Test Results
Assay | 99.8% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...