N-(2-Chloroethyl)benzamide ≥97%
Supplier: BeanTown Chemical
215590-10G
215590-50G
BT215590-10GEA
37
USD
BT215590-10G
BT215590-50G
N-(2-Chloroethyl)benzamide ≥97%
N-(2-Chloroethyl)benzamide
CAS: 26385-07-9; EC No: 247-641-4; MDL No: MFCD00000964
Crystalline/Powder; Molecular Formula: C9H10ClNO; MW: 183.64
Melting Point: 101-105°
- Crystalline/Powder
Formula:
C₉H₁₀ClNO Melting Pt: 101…105 °C |
MDL Number:
MFCD00000964 CAS Number: 26385-07-9 EINECS: 247-641-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...