2-Amino-6-chloropurine ≥99%

Supplier: BeanTown Chemical

Synonyms: 6-Chloroguanine, 6-Chloro-7H-purin-2-ylamine

215550-1G 215550-25G 215550-5G
BT215550-1GEA 58 USD
BT215550-1G BT215550-25G BT215550-5G
2-Amino-6-chloropurine ≥99%
2-Amino-6-chloropurine

CAS: 10310-21-1; EC No: 233-686-7; MDL No: MFCD00075252; RTECS: UO7502000 Powder; Molecular Formula: C5H4ClN5; MW: 169.57


  • RTECS: UO7502000
  • Powder

Formula: C₅H₄ClN₅
MW: 169.57 g/mol
MDL Number: MFCD00075252
CAS Number: 10310-21-1
EINECS: 233-686-7

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR