Thionin(acetate)
Supplier: BeanTown Chemical
|
Warning
|
Synonyms:
3,7-Diaminophenothiazin-5-ium acetate
215525-25G
215525-5G
BT215525-25GEA
152
USD
BT215525-25G
BT215525-5G
Thionin(acetate)
Thionin (Acetate)
CAS: 78338-22-4; MDL No: MFCD00081194
Solid; Linear Formula: C12H9N3S·C2H4O2; Molecular Formula: C14H13N3O2S; MW: 287.34
- Solid
Formula:
C₁₄H₁₃N₃O₂S MW: 287.34 g/mol |
MDL Number:
MFCD00081194 CAS Number: 78338-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...