Polyethyleneimine, M.W. ∼10000 ≥99%, branched

Supplier: BeanTown Chemical

Synonyms: Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)

215515-100G 215515-25G 215515-500G
BT215515-100GEA 132.51 USD
BT215515-100G BT215515-25G BT215515-500G
Polyethyleneimine, M.W. ∼10000 ≥99%, branched
Polyethyleneimine

CAS: 9002-98-6; MDL No: MFCD00803910; RTECS: TQ7915000 Liquid; Linear Formula: (-NHCH2CH2-)x[-N(CH2CH2NH2)CH2CH2-]y


  • RTECS: TQ7915000
  • Liquid

Formula: (CH2CH2NH)n
MDL Number: MFCD00084427
CAS Number: 9002-98-6

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR