Rifampicin
Supplier: BeanTown Chemical
Synonyms:
Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifamycin AMP
215500-1G
215500-25G
215500-5G
BT215500-1GEA
83.79
USD
BT215500-1G
BT215500-25G
BT215500-5G
Rifampicin
Rifampicin
CAS: 13292-46-1; EC No: 236-312-0; MDL No: MFCD00151389; RTECS: VJ7000000
Powder; Molecular Formula: C43H58N4O12; MW: 822.94
Melting Point: 194°
Light Sensitive, Moisture Sensitive
- Powder
- Light and moisture sensitive
Formula:
C₄₃H₅₈N₄O₁₂ MW: 822.95 g/mol Melting Pt: 194 °C |
MDL Number:
MFCD00151389 CAS Number: 13292-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...