Potassium formate ≥98%

Supplier: BeanTown Chemical

Synonyms: Formic acid potassium salt

215470-1KG 215470-250G 215470-5KG
BT215470-1KGEA 125.87 USD
BT215470-1KG BT215470-250G BT215470-5KG
Potassium formate ≥98%
Potassium formate

CAS: 590-29-4; EC No: 209-677-9; MDL No: MFCD00013100; RTECS: LQ9625000 Crystalline; Linear Formula: HCOOK; MW: 84.12 Melting Point: 165-168° Density (g/mL): 1.91 Hygroscopic


  • RTECS: LQ9625000
  • Hygroscopic
  • Crystalline

Formula: CHKO₂
MW: 84.12 g/mol
Melting Pt: 165…168 °C
Density: 1.91 g/mL
MDL Number: MFCD00013100
CAS Number: 590-29-4
EINECS: 209-677-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR