Creatinine ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one

215455-10G 215455-250G 215455-50G
BT215455-10GEA 26.61 USD
BT215455-10G BT215455-250G BT215455-50G
Creatinine ≥98%
Creatinine

CAS: 60-27-5; EC No: 200-466-7; MDL No: MFCD00059730 Solid; Molecular Formula: C4H7N3O; MW: 113.12 Melting Point: 295° (decomposes)


  • Solid

Formula: C₄H₇N₃O
MW: 113.12 g/mol
Melting Pt: 295 °C (dec.)
MDL Number: MFCD00059730
CAS Number: 60-27-5
EINECS: 200-466-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR