Dimethyl-2,5-pyridinedicarboxylate ≥98%

Supplier: BeanTown Chemical

Synonyms: Dimethyl pyridine-2,5-dicarboxylate

215450-1G 215450-25G 215450-5G
BT215450-1GEA 26 USD
BT215450-1G BT215450-25G BT215450-5G
Dimethyl-2,5-pyridinedicarboxylate ≥98%
Dimethyl 2,5-Pyridinedicarboxylate

CAS: 881-86-7; MDL No: MFCD00034767 Powder; Molecular Formula: C9H9NO4; MW: 195.17 Melting Point: 213-217°


  • Powder

Formula: C₉H₉NO₄
Melting Pt: 213…217 °C
MDL Number: MFCD00034767
CAS Number: 881-86-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR