4,5-Dichloroimidazole ≥98%

Supplier: BeanTown Chemical

Synonyms: 4,5-Dichloro-1H-imidazole

215435-25G 215435-5G
BT215435-25GEA 178 USD
BT215435-25G BT215435-5G
4,5-Dichloroimidazole ≥98%
4,5-Dichloroimidazole

CAS: 15965-30-7; MDL No: MFCD00005195 Powder; Molecular Formula: C3H2Cl2N2; MW: 136.97 Melting Point: 183-185°


  • Powder

Formula: C₃H₂Cl₂N₂
MW: 136.97 g/mol
Melting Pt: 183…185 °C
MDL Number: MFCD00005195
CAS Number: 15965-30-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR